Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB4700629

Sigma-Aldrich

Anti-VCP-FITC antibody

clone Hs-14, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Hs-14, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... VCP(7415)

Related Categories

General description

The antibody Hs-14 reacts with a 220 kDa testis-specific human intra-acrosomal protein associated with the membranes of the acrosomal vesicle.

Immunogen

Dialyzed acid extract of human sperm

Application

The reagent is designed for Flow Cytometry analysis using 20 μL reagent / 100 μL cell suspension. The content of a vial (1 mL) is sufficient for 50 tests.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service