Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700323

Sigma-Aldrich

Monoclonal Anti-HLA-G , (C-terminal) antibody produced in mouse

clone 5A6G7, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A6G7, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HLA-G(3135)

General description

Human histocompatibility leukocyte antigen–G (HLA-G) belongs to the non-classical class I HLA family. It is encoded by the gene mapped to human chromosome 6p21. Unlike other members of the human class I gene family, HLA-G is expressed specifically in extraembryonic tissues during gestation.
The antibody 5A6G7 allows to discriminate between soluble HLA-G protein yielded by shedding from membrane-bound HLA-G forms that do not contain the intron 4-encoded epitope and soluble HLA-G5/HLA-G6 produced from intron 4-retaining alternatively spliced mRNAs. HLA-G belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on the surface of trophoblast cells.

Immunogen

C-terminal amino acid sequence (22-mer) of soluble HLA-G5 and HLA-G6 proteins coupled to ovalbumin

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Human histocompatibility leukocyte antigen-G (HLA-G) molecule facilitates the regulation of maternal tolerance to the fetus by interacting with inhibitory receptors on natural killer (NK) cells. The encoded protein is involved in the embryonic development and/or the fetal-maternal immune response. Mutations in the gene increases the risk of susceptibility to a wide range of inflammatory diseases, including asthma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fine Mapping and Positional Candidate Studies Identify HLA-G as an Asthma Susceptibility Gene on Chromosome 6p21
Nicolae D
American Journal of Human Genetics, 76, 349-357 (2005)
Differential expression of HLA-E, HLA-F, and HLA-G transcripts in human tissue.
Wei XH and Orr HT
Human immunology, 29, 131-142 (1990)
A physical linkage map of HLA-A, -G, ?7.5p, and -F
Schmidt CM and Orr HT
Human immunology, 31, 180-185 (1991)
Recognition of human histocompatibility leukocyte antigen (HLA)-E complexed with HLA class I signal sequence-derived peptides by CD94/NKG2 confers protection from natural killer cell-mediated lysis.
Borrego F
The Journal of Experimental Medicine, 187, 813-818 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service