Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700079

Sigma-Aldrich

Monoclonal Anti-CD7 antibody produced in mouse

clone MEM-186, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-GP40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-186, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... CD7(924)

General description

The MEM-186 antibody reacts with CD7, a 40 kD type I transmembrane glycoprotein expressed on peripheral blood T lymphocytes, NK-cells, hematopoietic progenitors, monocytes (weakly) and also on acute lymphocytic leukemia.

Immunogen

Human acute myelogenous leukaemia cell line KG-1

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahul Chavan et al.
Cancer immunology research, 2(3), 241-248 (2014-04-30)
Monocytes may contribute to tumor progression in part by mediating tumor-induced immunosuppression. Alterations to the monocyte populations and functions in untreated patients with late-stage melanoma are not fully understood. To characterize these alterations, we compared the frequency, phenotype, and functional
Suchandra Chowdhury et al.
Glycoconjugate journal, 31(6-7), 523-535 (2014-10-07)
Childhood acute lymphoblastic leukaemia (ALL) originates from mutations in haematopoietic progenitor cells (HPCs). For high-risk patients, treated with intensified post-remission chemotherapy, haematopoietic stem cell (HSC) transplantation is considered. Autologous HSC transplantation needs improvisation till date. Previous studies established enhanced disease-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service