Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

SAB4504467

Sigma-Aldrich

Anti-phospho-MEK1/2 (pSer221) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
WB
clone:
polyclonal
species reactivity:
rat, mouse, human
citations:
2
technique(s):
ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer221)

Gene Information

Immunogen

The antiserum was produced against synthesized peptide derived from human MEK1/2 around the phosphorylation site of Ser221.

Immunogen Range: 193-242

Biochem/physiol Actions

MAP2k1/2 (dual specificity mitogen-activated protein kinase kinase 1/2) is a serine threonine kinase and is required for ERK (extracellular-signal-regulated kinase) activation. Activation of ERK is associated with production of IL-10 (interleukin 10) and IL-12. Absence of the Map2k1 gene results in embryonic lethality. Map2k1 is responsible for the stimulation of epidermal proliferation, regulating cell migration in fibroblasts. Deficiency of Map2k1 protein causes lupus-like syndrome. The gene encoding Map2k1 is localized on human chromosome 15q22.31 and the MAP2K2 gene is localized on human chromosome 19p13.3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MEK1 dependent and independent ERK activation regulates IL-10 and IL-12 production in bone marrow derived macrophages.
Bouhamdan M, et al.
Cellular Signalling, 27(10), 2068-2076 (2015)
The RASopathies: developmental syndromes of Ras/MAPK pathway dysregulation
Tidyman WE and Rauen KA
Current Opinion in Genetics & Development, 19(3), 230-236 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service