Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

SAB4504134

Sigma-Aldrich

Anti-phospho-CD4 (pSer433) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$428.00

$428.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$428.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$428.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 51 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer433)

Gene Information

human ... CD4(920)

General description

CD4 (CD4 molecule) is present in the surface of specific leukocytes. CD4 contains four ecto-domains (D1-D4) of which D1, D2, and D4 contain disulfide bonds. It is mapped to human chromosome 12p13.31. CD4 belongs to the immunoglobulin superfamily (IgSF). It has four extracellular Ig-like domains, a hydrophobic transmembrane region and a 40-residue cytoplasmic tail.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD4 around the phosphorylation site of Ser433.

Immunogen Range: 401-450

Biochem/physiol Actions

CD4 (CD4 molecule) helps in the activation of immunostimulatory T-cells. It serves as a primary receptor for HIV-1 entry. Individuals with infectious and chronic inflammatory diseases shows higher level of soluble CD4 (sCD4).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica De Santis et al.
Frontiers in immunology, 13, 830788-830788 (2022-06-07)
A strategy adopted to combat human immunodeficiency virus type-1 (HIV-1) infection is based on interfering with virus entry into target cells. In this study, we found that phosphatidylcholine (PC) liposomes reduced the expression of the CD4 receptor in human primary
Increased soluble CD4 in serum of rheumatoid arthritis patients is generated by matrix metalloproteinase (MMP)-like proteinases
Tseng WY, et al.
PLoS ONE (2013)
Human CD4 Metastability Is a Function of the Allosteric Disulfide Bond in Domain 2
Owen GR, et al.
Biochemistry, 55(15), 2227-2237 (2016)
Isochromosomes 12p and 9p: parental origin and possible mechanisms of formation
Dutly F, et al.
European Journal of Human Genetics, 6(2), 140-144 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service