Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4504011

Sigma-Aldrich

Anti-phospho-ZAP-70 (pTyr292) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 69 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr292)

Gene Information

human ... ZAP70(7535)

General description

Zeta-chain-associated protein kinase 70 (ZAP70) is a tyrosine kinase, present in T cells and natural killer (NK) cells. It belongs to the spleen tyrosine kinase (Syk) family. This gene is mapped to human chromosome 2q11.

Immunogen

The antiserum was produced against synthesized peptide derived from human ZAP-70 around the phosphorylation site of Tyr292.

Immunogen Range: 258-307

Biochem/physiol Actions

Zeta-chain-associated protein kinase 70 (ZAP70) plays an important role in T cell receptor (TCR) signaling. It acts as an adaptor protein to increase BCR (B-cell receptor) signaling. This protein is required for T-cell transendothelial migration mediated by C-X-C motif chemokine ligand 12.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An electrostatic selection mechanism controls sequential kinase signaling downstream of the T cell receptor.
Shah NH, et al.
eLife, 5, e20105-e20105 (2016)
The role of ZAP70 kinase in acute lymphoblastic leukemia infiltration into the central nervous system
Alsadeq A, et al.
Haematologica, 102(2), 346-355 (2017)
Mutations in Two Genes Encoding Different Subunits of a Receptor Signaling Complex Result in an Identical Disease Phenotype
Paloneva J, et al.
American Journal of Human Genetics, 71(3), 656-662 (2002)
Imaging Spatiotemporal Activities of ZAP-70 in Live T Cells Using a FRET-Based Biosensor
Li K, et al.
Annals of Biomedical Engineering, 44(12), 3510-3521 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service