Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4503974

Sigma-Aldrich

Anti-phospho-PLK1 (pThr210) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr210)

Gene Information

human ... PLK1(5347)

General description

Human polo-like kinase 1 (PLK1) is a serine/threonine kinase, that is located on human chromosome 16p12.

Immunogen

The antiserum was produced against synthesized peptide derived from human PLK1 around the phosphorylation site of Thr210.

Immunogen Range: 176-225

Biochem/physiol Actions

Polo-like kinase 1 (PLK1) plays an important role in the functional maturation of mitotic centrosomes. It actively participates in the development of the cell cycle. This gene is considered as an important therapeutic target as it is overexpressed in several cancer types.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drug sensitivity and resistance testing identifies PLK1 inhibitors and gemcitabine as potent drugs for malignant peripheral nerve sheath tumors
Kolberg M, et al.
Molecular Oncology, 11(9), 1156-1171 (2017)
Antibody microinjection reveals an essential role for human polo-like kinase 1 (Plk1) in the functional maturation of mitotic centrosomes
H A Lane and E A Nigg
The Journal of Cell Biology, 1701-1713 (1996)
Polo on the Rise-from Mitotic Entry to Cytokinesis with Plk1
Petronczki M, et al.
Developmental Cell, 14(5), 646-659 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service