Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4502810

Sigma-Aldrich

Anti-p47 PHOX antibody produced in rabbit

affinity isolated antibody

Synonym(s):

47 kDa autosomal chronic granulomatous disease protein, 47 kDa neutrophil oxidase factor, NCF-1, NCF-47K, NCF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$698.00

$698.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$698.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$698.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCF1(653361)

General description

Anti-p47 PHOX Antibody detects endogenous levels of total p47 PHOX protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human p47 phox.

Immunogen Range: 341-390

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Feillet-Coudray et al.
Lipids, 54(1), 81-97 (2019-02-16)
Inflammation and oxidative stress are thought to be involved in, or associated with, the development of obesity, dyslipidemia, hepatic steatosis, and insulin resistance. This work was designed to determine the evolution of inflammation and oxidative stress during onset and progression
Giulia Blandino et al.
Redox biology, 67, 102915-102915 (2023-10-23)
Long-term treatment of schizophrenia with clozapine (CLZ), an atypical antipsychotic drug, is associated with an increased incidence of metabolic disorders mediated by poorly understood mechanisms. We herein report that CLZ, while slowing down the morphological changes and lipid accumulation occurring
Kenta Kuramoto et al.
Molecular and cellular biology, 34(14), 2721-2731 (2014-05-14)
Lipid droplet (LD) is a ubiquitous organelle that stores triacylglycerol and other neutral lipids. Perilipin 5 (Plin5), a member of the perilipin protein family that is abundantly expressed in the heart, is essential to protect LDs from attack by lipases
Glaucy Rodrigues de Araújo et al.
Experimental biology and medicine (Maywood, N.J.), 242(3), 333-343 (2017-01-21)
Baccharis trimera, popularly known as "carqueja", is a native South-American plant possessing a high concentration of polyphenolic compounds and therefore high antioxidant potential. Despite the antioxidant potential described for B. trimera, there are no reports concerning the signaling pathways involved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service