Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

SAB4502202

Anti-PLCB3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase β 3, PLC-β-3, PLCB3, Phosphoinositide phospholipase C, Phospholipase C-β-3

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, WB
Citations:
-

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 138 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLCB3(5331)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4502203SAB5700499SAB4502206
conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

UniProt accession no.

Q01970

UniProt accession no.

Q01970

UniProt accession no.

Q01970

UniProt accession no.

P19174

technique(s)

ELISA: 1:40000, western blot: 1:500-1:1000, immunohistochemistry: 1:50-1:100

technique(s)

ELISA: 1:1000, immunohistochemistry: 1:50-1:100

technique(s)

western blot: 1:500-1:2000

technique(s)

ELISA: 1:1000, immunofluorescence: 1:100-1:500

General description

Anti-PLCB3 Antibody detects endogenous levels of total PLCB3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PLCB3.

Immunogen Range: 1071-1120

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
SAB4502202-100UG04061837174964

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service