Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4502130

Sigma-Aldrich

Anti-PRKAG2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

5′-AMP-activated protein kinase subunit γ-2, AAKG2, AMPK γ-2 chain, AMPK γ2, H91620p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 63 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKAG2(51422)

General description

Anti-PRKAG2 Antibody detects endogenous levels of total PRKAG2 protein.
PRKAG2 (protein kinase, AMP-activated, γ2 non-catalytic subunit) is a non-catalytic regulatory subunit of heterotrimeric AMP-activated protein kinase (AMPK), involved in the energy metabolism. It is predominantly present in all subcellular fractions, including the nuclei.

Immunogen

The antiserum was produced against synthesized peptide derived from human PRKAG2.

Immunogen Range: 1-50

Application

Anti-PRKAG2, N-Terminal antibody produced in rabbit is suitable for immunohistochemistry and indirect immunofluorescence.

Biochem/physiol Actions

PRKAG2 (protein kinase, AMP-activated, γ2 non-catalytic subunit) acts as a sensor of cellular energy status that is controlled by AMP (positively) and ATP (negatively). In normal state CBS domains from AMP-activated protein kinase can bind AMP, ATP, or S-adenosyl methionine. But in mutated condition, it cannot bind. As a result, it leads to hereditary diseases. Its major activity is involved in the regulation of energy homeostasis. It plays an important role in correcting of cell division and chromosome segregation during mitosis. Mutation in PRKAG2 causes Wolff-Parkinson-White (WPW) syndrome, a rare autosomal dominant inheritance disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katalin Pinter et al.
Cell cycle (Georgetown, Tex.), 11(5), 917-921 (2012-02-16)
AMP-activated protein kinase has been shown to be a key regulator of energy homeostasis; it has also been identified as a tumor suppressor and is required for correct cell division and chromosome segregation during mitosis. The enzyme is a heterotrimer
Tània Martiáñez et al.
The Journal of biological chemistry, 284(36), 23902-23911 (2009-07-03)
Ultrasensitivity, hysteresis (a form of biochemical memory), and all-or-none (digital) responses are important signaling properties for the control of irreversible processes and are well characterized in the c-Jun N-terminal kinase (JNK) system using Xenopus oocytes. Our aim was to study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service