Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4502007

Sigma-Aldrich

Anti-NKTR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NK-TR protein, NK-tumor recognition protein, Natural-killer cells cyclophilin-related protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 165 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKTR(4820)

Related Categories

General description

NKTR (Natural killer-tumor recognition sequence) is a 150kDa subunit of a putative tumor-recognition complex. It is expressed on the surface of natural killer cells. It contains an amino-terminal hydrophobic region, a cyclophilin-related domain, three histone-like domains, and three arginine and serine-rich domains.

Immunogen

The antiserum was produced against synthesized peptide derived from human NKTR.

Immunogen Range: 784-833

Application

Anti-NKTR antibody produced in rabbit is suitable for immunohistochemistry and indirect immunofluorescence studies.

Biochem/physiol Actions

NKTR (Natural killer-tumor recognition sequence) plays an important role in NK-cell cytotoxicity in human T-cells. It participates in identifying as well as NK-like killing. It has been reported that IL2 (interleukin 2) control the NKTR activity by altering the RNA splicing pattern.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Anderson et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(2), 542-546 (1993-01-15)
Natural killer cells are non-major histocompatibility complex-restricted large granular lymphocytes that can recognize and destroy tumor cells without prior stimulation. A 150-kDa molecule on the surface of human natural killer cells was identified as a component of a putative tumor-recognition
C A Chambers et al.
Journal of immunology (Baltimore, Md. : 1950), 152(6), 2669-2674 (1994-03-15)
NK cells lyse target cells without previous immune sensitization. A small subset of T cells also exhibits NK-like activity, which is distinct from TCR-mediated, MHC-restricted, and MHC-unrestricted cytotoxicity. We recently cloned a gene, NK-TR, which is postulated to be part
A Rinfret et al.
Molecular immunology, 30(14), 1307-1313 (1993-10-01)
We have recently isolated and characterized human and mouse genes of a putative natural killer (NK) cell tumour-recognition protein (NK-TR) that is specifically expressed in NK cells. This gene codes for a 150 kD protein with a cyclophilin-related amino terminus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service