Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB4501647

Sigma-Aldrich

Anti-KIT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD117 antigen, Proto-oncogene tyrosine-protein kinase Kit, SCFR, c-kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 109 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KIT(3815)

General description

Anti-KIT Antibody detects endogenous levels of total KIT protein.
KIT (proto-oncogene) is a type III receptor tyrosine kinase (RTK). It codes for a tyrosine kinase receptor (CD117). It is located on human chromosome 4q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human KIT.

Immunogen Range: 906-955

Biochem/physiol Actions

KIT (proto-oncogene) participates in cell signal transduction. It also participates in the progression of hematopoietic stem cells, melanocytes, germ cells, mast cells and interstitial cells of Cajal. Mutations in KIT gene results in gastrointestinal stromal tumors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Wang et al.
Cell communication and signaling : CCS, 16(1), 16-16 (2018-04-18)
Gain-of-function mutations and overexpression of KIT are characteristic features of gastrointestinal stromal tumor (GIST). Dysregulation in miRNA expression may lead to KIT overexpression and tumorigenesis. miRNA microarray analysis and real-time PCR were used to determine the miRNA expression profiles in
Loss of c-KIT expression in thyroid cancer cells.
Franceschi S, et al.
PLoS ONE, 12(3) (2017)
KIT extracellular and kinase domain mutations in gastrointestinal stromal tumors
Lux ML, et al.
The American Journal of Pathology, 156(3), 791-795 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service