Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4501435

Sigma-Aldrich

Anti-APBB2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Amyloid β A4 precursor protein-binding family B member 2, Fe65-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 83 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APBB2(323)

General description

The gene APBB2 (amyloid β precursor protein binding family B member 2) is mapped to human chromosome 4p13.

Immunogen

The antiserum was produced against synthesized peptide derived from human APBB2.

Immunogen Range: 471-520

Biochem/physiol Actions

The gene APBB2 (amyloid β precursor protein binding family B member 2), also referred to as FE65-like 1, encodes an adaptor protein that binds to the cytoplasmic domain of β-amyloid precursor protein (βAPP) and processes it to form β-amyloid (Aβ). Over-expression of APBB2 leads to an accumulation of Aβ in senile plaques seen in patients with Alzheimer′s disease (AD). Mutations in this gene have been associated with AD.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zfra is an inhibitor of Bcl-2 expression and cytochrome c release from the mitochondria.
Hsu LJ
Cellular Signalling, 20, 1303-1312 (2008)
APBB2 genetic polymorphisms are associated with severe cognitive impairment in centenarians.
Golanska E
Experimental Gerontology, 48, 391-394 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service