Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300995

Sigma-Aldrich

Anti-HSD17B13 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HMFN0376, NIIL497, SCDR9, SDR16C3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$440.00

$440.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$440.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$440.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

human

concentration

1.3 mg/mL

technique(s)

immunohistochemistry: 1:50- 1:200
western blot: 1:500-1:2000 (Cell Lysate)

isotype

IgG

accession no.

NP_001129702.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Hydroxysteroid 17-beta dehydrogenase 13 (HSD17B13) is part of the short-chain dehydrogenase/reductase family. The gene encoding this lipid droplet-associated protein is localized on human chromosome 4q22.1.

Specificity

The antibody detects endogenous levels of total HSD17B13 protein.

Immunogen

Fusion protein corresponding to a region derived from internal residues of human hydroxysteroid (17-beta) dehydrogenase 13

Biochem/physiol Actions

Hydroxysteroid 17-beta dehydrogenase 13 (HSD17B13) has a role in non-alcoholic fatty liver disease (NAFLD) and it has been shown to be downregulated in hepatocellular carcinoma. HSD17B13 modulates lipid biosynthesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service