Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300689

Sigma-Aldrich

Anti-ABCG2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ABC15, Anti-ABCP, Anti-BMDP, Anti-CD338, Anti-MRX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65-80 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-I-N-R-V)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCG2(9429)

General description

The ABCG2 (ATP binding cassette subfamily G member 2) gene encodes an ATP-binding cassette (ABC) transporter that is also called as breast cancer resistance protein (BRCP). It is composed of 655 amino-acids and is located on human chromosome 4q22. It belongs to the ABCG (white) subfamily.

The antibody detects endogenous level of total ABCG2(CD338) protein.

Immunogen

Peptide sequence around aa. 160-164 ( R-I-N-R-V), according to the protein NP_004818.2

Biochem/physiol Actions

ABCG2 (ATP binding cassette subfamily G member 2) serves as a shield for several tissues against xenobiotic molecules. It can provide multidrug resistance (MDR) in breast and other cell lines.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Xenobiotic transporter that may play an important role in the exclusion of xenobiotics from the brain. May be involved in brain-to-blood efflux. Appears to play a major role in the multidrug resistance phenotype of several cancer cell lines. When overexpressed, the transfected cells become resistant to mitoxantrone, daunorubicin and doxorubicin, display diminished intracellular accumulation of daunorubicin, and manifest an ATP-dependent increase in the efflux of rhodamine 123.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multidrug resistance mediated by the breast cancer resistance protein BCRP (ABCG2)
Doyle L and Ross DD
Oncogene (2003)
Structure of the human multidrug transporter ABCG2
Taylor NMI, et al.
Nature (2017)
Kinetic Modeling of ABCG2 Transporter Heterogeneity: A Quantitative, Single-Cell Analysis of the Side Population Assay
Prasanphanich AF, et al.
PLoS Computational Biology (2016)
Gayathri Heenatigala Palliyage et al.
Lung cancer (Amsterdam, Netherlands), 181, 107258-107258 (2023-05-29)
A cure for cancer is out of reach for most patients due to chemoresistance. Cancer-associated fibroblasts (CAFs) play a vital role in cancer chemoresistance, but detailed understanding of the process particularly in chemoresistant lung cancer is lacking. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service