Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300454

Sigma-Aldrich

Anti-EGFR (Ab-693) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ERBB antibody produced in rabbit, Anti-ERBB1 antibody produced in rabbit, Anti-HER1 antibody produced in rabbit, Anti-PIG61 antibody produced in rabbit, Anti-epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~175 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-L-T-P-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)

General description

Epidermal growth factor receptor (EGFR, ErbB1) is a transmembrane protein that exerts tyrosine kinase activity upon ligand induced activation. EGFR can be activated by binding EGF or at least six other structurally related protein ligands, including transforming growth factor-α (TGF-α), heparin-binding EGF-like growth factor (HB-EGF), betacellulin (BTC), amphiregulin, epiregulin and epigen. The gene encoding it is localized on human chromosome 7p11.2.

Immunogen

Peptide sequence around aa. 691-695 (P-L-T-P-S), according to the protein EGFR.

Biochem/physiol Actions

Upon activation by ligands, epidermal growth factor receptor (EGFR, ErbB1) initiates a signaling cascade which includes dimerization and internalization, tyrosine phosphorylation, DNA synthesis of target genes, and, ultimately, cell proliferation. EGFR signaling plays a role in the growth and differentiation of normal cells, but elevated EGFR activity is correlated with the development and pathogenesis of certain cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for EGF, but also for other members of the EGF family, as TGF-alpha, amphiregulin, betacellulin, heparin-binding EGF-like growth factor, GP30 and vaccinia virus growth factor. Is involved in the control of cell growth and differentiation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A chromosomal region 7p11.2 transcript map: its development and application to the study of EGFR amplicons in glioblastoma.
Eley GD
Neuro-Oncology, 4(2), 86-94 (2002)
Molecular mechanisms of resistance to the EGFR monoclonal antibody cetuximab.
Brand TM
Cancer Biology & Therapy, 11(9), 777-792 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service