Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300342

Sigma-Aldrich

Anti-BAD (Ab-136) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BBC2 antibody produced in rabbit, Anti-BCL2-associated agonist of cell death antibody produced in rabbit, Anti-BCL2L8 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~23 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(S-R-S-A-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAD(572)

General description

BAD (BCL2 associated agonist of cell death) is a proapoptotic protein. It is a 184 kDa protein made of 168 amino acids. It belongs to the Bcl2 family of proteins. The gene encoding it is localized on human chromosome 11q13.

Immunogen

Peptide sequence around aa. 134-138 (S-R-S-A-P), according to the protein BAD.

Biochem/physiol Actions

Phosphorylated BAD (BCL2 associated agonist of cell death) has the ability to interact with 14-3-3 scaffold proteins in the cytoplasm. Dephosphorylated BAD is active and it moves to the outer membrane of mitochondria and make heterodimers with BCL-XL to prevent BCL-XL antiapoptotic activities.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The protein encoded by BAD gene is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BAD (BCL2-antagonist of cell death).
Huret JL and Senon S
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service