Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300333

Sigma-Aldrich

Anti-CREB1 (Ab-133) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CREB antibody produced in rabbit, Anti-MGC9284 antibody produced in rabbit, Anti-cAMP responsive element binding protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~43 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-P-S-Y-R)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)

General description

CREB1 (cAMP response element-binding protein) gene codes for a 43kDa cellular transcription factor, CREB. It has 341 amino acids. It belongs to the basic leucine zipper family of transcription factors. It is upregulated abundantly in human tissues and is located on chromosome 2q33–35.

Immunogen

Peptide sequence around aa. 131-135 (R-P-S-Y-R), according to the protein CREB1.

Biochem/physiol Actions

Knockdown of CREB (cAMP response element-binding protein) blocks the development and promotes apoptosis in pre-B acute lymphoblastic leukemia cells by preventing prosurvival signals. It controls the transcription of antiapoptotic proteins in several cell types. Decrease in CREB phosphorylation leads to malfunction of mitochondria and hypoxia-induced apoptosis in chondrocytes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

This protein binds the cAMP response element (CRE), a sequence present in many viral and cellular promoters. CREB stimulates transcription on binding to the CRE. Transcription activation is enhanced by the TORC coactivators which act independently of Ser-133 phosphorylation. Implicated in synchronization of circadian rhythmicity.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Varicella-Zoster Virus Activates CREB, and Inhibition of the pCREB-p300/CBP Interaction Inhibits Viral Replication In Vitro and Skin Pathogenesis In Vivo
Francois S, et al.
Journal of Virology (2016)
CREB knockdown inhibits growth and induces apoptosis in human pre-B acute lymphoblastic leukemia cells through inhibition of prosurvival signals
Shabestari RM, et al.
Biomedicine and Pharmacotherapy (2017)
Hypoxia-induced apoptosis and mitochondrial dysfunction in chondrocytes arising from CREB phosphorylation reduction
Qiu YY, et al.
Genetics and molecular research : GMR (2016)
Oxidative stress-induced CREB upregulation promotes DNA damage repair prior to neuronal cell death protection
Pregi N, et al.
Molecular and Cellular Biochemistry (2017)
Effects of the G(-656)A variant on CREB1 promoter activity in a neuronal cell line: interactions with gonadal steroids and stress
Zubenko GS and Hughes HB
Molecular Psychiatry (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service