Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300328

Sigma-Aldrich

Anti-STAT4 (Ab-693) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-SLEB11 antibody produced in rabbit, Anti-signal transducer and activator of transcription 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~85 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(K-G-Y-V-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT4(6775)

Immunogen

Peptide sequence around aa. 691-695 (K-G-Y-V-P), according to the protein STAT4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Carries out a dual function: signal transduction and activation of transcription. Involved in IL12 signaling.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Lv et al.
Journal of molecular and cellular cardiology, 74, 284-294 (2014-06-17)
Signal transducer and activator of transcription 4 (STAT4) has been associated with susceptibility to autoimmune diseases. Intriguingly, we previously reported that STAT4 might play a critical role in vascular smooth muscle cell (VSMC) proliferation. The present study therefore investigated the
Xiaoying Zhou et al.
Helicobacter, 19(3), 174-181 (2014-03-19)
Recent studies found that gastric cancer patients with Helicobacter pylori infection had a better response to chemotherapy and had an improved overall prognosis compared with those without. However, the underlying mechanism remains unknown. Quantitative real-time PCR (qRT-PCR) was utilized to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service