Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300206

Sigma-Aldrich

Anti-phospho-GJA1 (pSer368) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CX43 antibody produced in rabbit, Anti-DFNB38 antibody produced in rabbit, Anti-GJAL antibody produced in rabbit, Anti-ODDD antibody produced in rabbit, Anti-gap junction protein, alpha 1, 43kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~43 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-A-SP-S-R)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer368)

Gene Information

human ... GJA1(2697)

Immunogen

Peptide sequence around phosphorylation site of serine 368 (R-A-S(p)-S-R), according to the protein GJA1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell. May play a critical role in the physiology of hearing by participating in the recycling of potassium to the cochlear endolymph.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenying Wang et al.
Experimental and therapeutic medicine, 17(3), 1847-1854 (2019-02-21)
Cx43 has been documented to be involved in ischemic preconditioning (IPC). However, the participation of Cx43-formed hemichannels in IPC and the potential underlying mechanisms remain unclear. The present study focused on cardiomyocytes' volume regulation during IPC to investigate the role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service