Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB4300059

Sigma-Aldrich

Anti-phospho-MYOD1 (pSer200) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MYF3 antibody produced in rabbit, Anti-MYOD antibody produced in rabbit, Anti-PUM antibody produced in rabbit, Anti-bHLHc1 antibody produced in rabbit, Anti-myogenic differentiation 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~40 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(A-S-SP-P-R)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer200)

Gene Information

human ... MYOD1(4654)

Immunogen

Peptide sequence around phosphorylation site of serine 200 (A-S-S(p)-P-R), according to the protein MYOD1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

MyoD encodes a nuclear protein that belongs to the basic helix-loop-helix family of transcription factors and themyogenic factors subfamily. It regulates muscle cell differentiation by inducing cell cycle arrest, a prerequisite formyogenic initiation. The protein is also involved in muscle regeneration. It activates its own transcription which maystabilize commitment to myogenesis.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service