Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4300056

Sigma-Aldrich

Anti-phospho-PGR (pSer190) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NR3C3 antibody produced in rabbit, Anti-PR antibody produced in rabbit, Anti-progesterone receptor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~99 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(G-L-SP-P-A)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer190)

Gene Information

human ... PGR(5241)

Related Categories

Immunogen

Peptide sequence around phosphorylation site of serine 190 (G-L-S(p)-P-A), according to the protein PGR.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Progesterone receptors (PRs) are nuclear hormone receptors of the NR3C class, which also includes mineralocorticoid,glucocorticoid and androgen receptors. They exist as homodimers coupled to Hsp90 or HMGB proteins, which are shed upon activation. The major signaling pathway used by progesterone receptors is via direct DNA binding and transcriptional regulation of target genes. They can also signal by binding to other proteins, mainly with transcription factors such as NF-kappaB, AP-1 or STAT. Progesterone receptors are found in the female reproductive tract, mammary glands, brain and pituitary gland and receptor expression is induced by estrogen. Well established functions of progesterone receptors include ovulation, implantation, mammary gland development and maintenance of pregnancy. In addition,progesterone, signaling through the progesterone receptor, increases the ventilatory response of the respiratory centers to carbon dioxide and decreases arterial and alveolar PCO2 in the luteal phase of the menstrual cycle and during pregnancy. The human gene encoding the progesterone receptor has been localized to 11q22.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service