Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300045

Sigma-Aldrich

Anti-phospho-NBN (pSer343) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AT-V1 antibody produced in rabbit, Anti-AT-V2 antibody produced in rabbit, Anti-ATV antibody produced in rabbit, Anti-FLJ10155 antibody produced in rabbit, Anti-nibrin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~95 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(S-L-SP-Q-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer343)

Gene Information

human ... NBN(4683)

Immunogen

Peptide sequence around phosphorylation site of serine 343 (S-L-S(p)-Q-G), according to the protein NBN.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Mutations in p95/NBS1 gene are associated with Nijmegen breakage syndrome, an autosomal recessive chromosomal instability syndrome characterized by microcephaly, growth retardation, immunodeficiency, and cancer predisposition. The encoded protein is a member of the MRE11/RAD50 double-strand break repair complex which consists of 5 proteins. This gene product is thought to be involved in DNA double-strand break repair and DNA damage-induced checkpoint activation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service