Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300010

Sigma-Aldrich

Anti-phospho-REL (pSer503) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C-Rel antibody produced in rabbit, Anti-v-rel reticuloendotheliosis viral oncogene homolog (avian) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
clone:
polyclonal
species reactivity:
human
citations:
technique(s):
indirect immunofluorescence: 1:100-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~78 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(T-S-SP-D-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer503)

Gene Information

human ... REL(5966)

Immunogen

Peptide sequence around phosphorylation site of serine 503 (T-S-S(p)-D-S), according to the protein REL.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA(MIM 164014), RELB (604758), NFKB1 (MIM 164011), and NFKB2 (MIM 164012). These proteins are related through a highlyconserved N-terminal region termed the 'Rel domain,' which is responsible for DNA binding, dimerization,nuclear localization, and binding to the NFKB inhibitor (MIM 164008).

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service