Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200827

Sigma-Aldrich

Anti- Staphylococcal alpha-Toxin-Peroxidase antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Alpha-hemolysin (hly)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

Research area: Microbiome

form

lyophilized powder

mol wt

~33 kDa

species reactivity

Staphylococcus aureus (alpha-Toxin (a-Hemolysin) )

concentration

~4 mg/mL

technique(s)

direct ELISA: 1:2,000-1:4,000 using 1 μg/mL Staphylococcal alpha-Toxin (a-Hemolysin) for coating.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Staphylococcus aureus subsp. aureus NCTC 8325 ... SAOUHSC_01121(3920722)

General description

Staphylococcal alpha-Toxin is a pore forming cytotoxin. Native alpha toxin is a water-soluble monomer but later on gets converted to β-barrel in the host membranes. It is 34 kDa protein. The N- terminal and the C-terminal are involved in forming a membrane-binding epitope and the central domain forms a membrane-inserting, pore forming domain.

Specificity

Anti-Staphylococcal alpha-Toxin-peroxidase antibody specifically recognizes Staphylococcal alpha-Toxin (?-Hemolysin) and has no cross reactivity with Staphylococcal Enterotoxin A (SEA), Cholera Toxin or Pseudomonas Exotoxin A

Immunogen

purified toxin from Staphylococcus aureus

Application

The antibody may be used in various immunochemical techniques including ELISA

Biochem/physiol Actions

Staphylococcal alpha-Toxin plays a role in binding to the host membrane. At higher concentrations it makes the cells which are devoid of receptors to become more permeable and hence also to protein-free lipid bilayers.

Diagnostic Implications

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Physical form

Supplied as a lyophilized powder.

Storage and Stability

Store the lyophilized product at 2–8 °C. For extended storage after reconstitution, keep at –20 °C in working aliquots. Avoid repeated freeze-thaw cycles. For continuous use after reconstitution, keep at 2–8 °C for up to 1 month. Solutions at working dilution should be discarded if not used within 12 hours.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Staphylococcal alpha-toxin, streptolysin-O, and Escherichia coli hemolysin: prototypes of pore-forming bacterial cytolysins
Bhakdi S, et al.
Archives of Microbiology, 165(2), 73-79 (1996)
Role of a disintegrin and metalloprotease 10 in Staphylococcus aureus alpha-hemolysin-mediated cellular injury
Wilke GA and Wardenburg JB
Proceedings of the National Academy of Sciences of the USA, 107(30), 13473-13478 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service