Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB4200738

Sigma-Aldrich

Anti-FITC antibody, Mouse monoclonal

clone FL-D6, purified from hybridoma cell culture

Synonym(s):

Anti-Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

FL-D6, monoclonal

form

buffered aqueous solution

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 1.25-2.5 μg/mL using ExtrAvidin-FITC
immunohistochemistry: suitable
indirect ELISA: 0.15-0.3 μg/mL using 5 μg/ml ExtrAvidin-FITC

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-FITC antibody, Mouse monoclonal, (mouse IgG1 isotype) is derived from the FL-D6 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with FITC (fluorescein isothiocyanate)-BSA conjugate. This antibody specifically recognizes both free and conjugated FITC.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light (with peak wavelength of approximately 495 nm) causing molecules to become excited and emit a visible yellow-green light (with peak wavelength of approximately 519 nm).

Specificity

Monoclonal Anti-FITC antibody specifically recognizes both free and conjugated FITC. The antibody does not react with bound or free TRITC (tetramethylrhodamine isothiocyanate).

Immunogen

FITC-BSA conjugate

Application

Anti-FITC antibody, Mouse monoclonal may be used in:
  • immunoblotting
  • enzyme-linked immunosorbent assay (ELISA)
  • immunohistochemistry

Biochem/physiol Actions

FITC (fluorescein isothiocyanate) is a commonly used protein marker including antibodies in immunofluorescent techniques. Antibodies to FITC are used to identify FITC labeled proteins and as models to study the mechanism of antibody response to a well-defined hapten.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20°C. For continuous use, store at 2–8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation. Discard working dilution samples if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elsa Charignon et al.
International journal of molecular sciences, 21(21) (2020-10-28)
We describe here the evaluation of the cytotoxic efficacy of two platinum (II) complexes bearing an N-heterocyclic carbene (NHC) ligand, a pyridine ligand and bromide or iodide ligands on a panel of human metastatic cutaneous melanoma cell lines representing different
Supriya Sharma et al.
Diagnostics (Basel, Switzerland), 12(1) (2022-01-22)
Isothermal techniques with lateral flow detection have emerged as a point of care (POC) technique for malaria, a major parasitic disease in tropical countries such as India. Plasmodium falciparum and Plasmodium vivax are the two most prevalent malaria species found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service