Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200654

Sigma-Aldrich

Monoclonal Anti-CHD1 antibody produced in rat

clone 2F11H5, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F11H5, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using using HeLa cells
immunofluorescence: 20 μg/mL using using human HeLa cells.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHD1(1105)

General description

Monoclonal Anti-CHD1 (rat IgG2a isotype) is derived from the hybridoma 2F11H5 produced by the fusion of mouse myeloma cells and lymph node cells from rat immunized with a synthetic peptide. Chromodomain-helicase-DNA-binding protein 1 (CHD1) is an ATP-dependent chromatin-remodeling factor expressed majorly in cerebellum and basal ganglia. CHD1 gene is mapped to human chromosome 5q15-q21.1. CHD1 comprises two chromodomains.

Specificity

Monoclonal Anti-CHD1 recognizes human CHD1.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminal region of human CHD1.

Application

Monoclonal Anti-CHD1 antibody produced in rat may be used in:
  • immunofluorescence
  • flow cytometry
  • immunoblotting
  • immunohistochemistry

Biochem/physiol Actions

Chromodomain-helicase-DNA-binding protein 1 (CHD1) interacts with the trimethylated lysine residues of histones and alters access of the transcriptional apparatus to its chromosomal DNA template. CHD1 is required for maintenance of open chromatin and pluripotency in embryonic stem cells. Missense mutation in the CHD1 gene is implicated in the Pilarowski-Bjornsson syndrome as well as in speech delay and facial dismorphism.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage,freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Familial segregation of a 5q15-q21. 2 deletion associated with facial dysmorphism and speech delay
Zepeda-Mendoza
Clinical case reports, 7(6), 1154-1160 (2019)
Missense variants in the chromatin remodeler CHD1 are associated with neurodevelopmental disability
Pilarowski GO, et al.
Journal of medical Genetics, 55(8), 561-566 (2018)
Lena Ho et al.
Nature, 463(7280), 474-484 (2010-01-30)
New methods for the genome-wide analysis of chromatin are providing insight into its roles in development and their underlying mechanisms. Current studies indicate that chromatin is dynamic, with its structure and its histone modifications undergoing global changes during transitions in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service