Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200480

Sigma-Aldrich

Anti-Kindlin-1 antibody, Mouse monoclonal

clone 4A5.14, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-C20orf42, Monoclonal Anti-DTGCU2, Monoclonal Anti-FERMT1 URP1, Monoclonal Anti-KIND1, Monoclonal Anti-Kindlin-1 antibody produced in mouse, Monoclonal Anti-UNC112A, Monoclonal Anti-fermitin family member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

4A5.14, monoclonal

form

buffered aqueous solution

mol wt

antigen ~77 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: 2.5-5.0 μg using SW-48 cells
immunoprecipitation (IP): suitable
western blot: 2.0-4.0 μg/mL using SW-48 total cell extracts

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FERMT1(55612)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Kindlin-1 (mouse IgG2a isotype) is derived from the hybridoma 4A5.14 produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Kindlin-1 is a focal adhesion protein, which belongs to the family of Kindlin proteins. It is highly expressed in epithelial cells, including keratinocytes. Kindlin-1 is localized to the focal adhesions. A splice variant potentially producing a truncated protein has been detected in the kidney, colon and small intestine. Kindlin-1 comprises a bipartite 4.1 protein, ezrin, radixin, moesin (FERM) domain with four subdomains (F0, F1, F2, and F3), which is found in many proteins. The KIND1 gene is present at human chromosome location 20p12.3.

Specificity

Monoclonal Anti-Kindlin-1 recognizes human Kindlin-1.

Immunogen

human Kindlin-1 recombinant fusion protein.

Application

Monoclonal Anti-Kindlin-1 antibody produced in mouse may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry
  • flow cytometry

Biochem/physiol Actions

Kindlin-1 is implicated in the activation of integrin adhesion receptors and the reorganization of the actin cytoskeleton. Deficiency of kindlin-1 is linked to Kindler syndrome, an autosomal recessive genodermatosis characterized by skin blistering, progressive skin atrophy, photosensitivity and carcinogenesis. This gene is also implicated in breast cancer, lung metastasis and lung tumorigenesis. The 4.1 protein, ezrin, radixin, moesin (FERM) domain of Kindlin-1 is involved in cytoskeletal organization. In colon cancer, Kindlin-1 is identified as a potential marker. Kindlin-1 modulates epidermal growth factor receptor signaling.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kindlin-1 expression is involved in migration and invasion of pancreatic cancer
Mahawithitwong P, et al.
International Journal of Oncology, 42(4), 1360-1366 (2013)
Loss of kindlin-1, a human homolog of the Caenorhabditis elegans actin-extracellular-matrix linker protein UNC-112, causes Kindler syndrome
Siegel DH, et al.
American Journal of Human Genetics, 73(1), 174-187 (2003)
Kindlin-1 Regulates Epidermal Growth Factor Receptor Signaling
Michael M, et al.
The Journal of Investigative Dermatology, 139(2), 369-379 (2019)
Kindlins in FERM adhesion
Malinin NL, et al.
Blood, 115(20), 4011-4017 (2010)
D J Papachristou et al.
Histopathology, 51(4), 499-508 (2007-08-23)
Leiomyosarcomas (LMS) are malignant neoplasms composed of cells that exhibit distinct smooth muscle differentiation. The molecular and cytogenetic features of LMS are complex and no consistent aberrations have been reported to date. Mitogen inducible gene-2 (Mig-2), kindlin and migfilin are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service