Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200439

Sigma-Aldrich

Anti-EXOSC5 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Exosome component 5, p12B, Anti-RRP41B, Anti-RRP46, Anti-Rrp46p, Anti-hRrp46p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~25 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using lysates of human HeLa cells.
indirect immunofluorescence: 1-2 μg/mL using human HeLa cells.
western blot: 1-2 μg/mL using whole extracts of human HEK-293T cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EXOSC5(56915)

General description

Exosome component 5 (EXOSC5) protein is a non-catalytic element of the RNA exosome complex in eukaryotes. The EXOSC5 gene is located on the human chromosome at 19q13.2.

Specificity

Anti-EXOSC5 (N-terminal) recognizes human EXOSC5.

Immunogen

peptide corresponding to the N-terminal region of human EXOSC5, conjugated to KLH. The antibody is affinity-purified using the immunizing peptide immobilized on agarose.

Application

Anti-EXOSC5 (N-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Exosome component 5 (EXOSC5) plays a role in the processing and degradation of RNA. This protein also plays a role in cleaving the DNA during apoptosis. EXOSC5 possesses a 3′-5′ exoribonuclease activity. Overexpression of the EXOSC5 gene is associated with several cancers such as melanoma, prostate, colorectal and lung cancer.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. Forextended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongda Pan et al.
Frontiers in oncology, 9, 643-643 (2019-08-06)
Background and Objective: Exosome component 5 (EXOSC5) is a novel cancer-related gene that is aberrantly expressed in various malignances. However, the molecular mechanism and biological role of EXOSC5 have not been explored in colorectal cancer (CRC). In this study, we
Jillian de Wilde et al.
International journal of cancer, 122(4), 877-888 (2007-10-26)
Although high-risk human papillomavirus (HPV) infection plays a major role in the development of cervical cancer, additive oncogenic events are involved as well. One key event involves increased activity of telomerase resulting from a deregulated expression of its catalytic subunit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service