Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB4200424

Sigma-Aldrich

Anti-COG1 (C-TERMINAL) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CDG2G, Anti-COG complex subunit 1, Anti-LDLB, Anti-component of oligomeric golgi complex 1, Anti-conserved oligomeric Golgi complex subunit 1, Anti-low density lipoprotein receptor defect

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

mouse, human, rat

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 1-2 μg/mL using human HeLa cells.
western blot: 2.5-5.0 μg/mL using whole extracts of mouse LA-4 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COG1(9382)
mouse ... Cog1(16834)
rat ... Cog1(303652)

Related Categories

General description

COG1 is a member of the conserved oligomeric Golgi (COG) complex. COG complex is an evolutionarily conserved multi-subunit protein complex. COG complex consists of eight distinct subunits organized in two heterotrimeric groups, Cog2-Cog3-Cog4 and Cog5-Cog6-Cog7, which are linked by the dimeric group formed by Cog1 and Cog8.
Conserved oligomeric Golgi (COG) subunit 1 is an essential component of the conserved oligomeric Golgi complex. It is encoded by the gene mapped to human chromosome 17q25.1.

Immunogen

peptide corresponding to the C-terminal region of human COG1, conjugated to KLH. The corresponding sequence is identical in mouse, rat and monkey COG1.

Application

Anti-COG1 (C-TERMINAL) antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

COG1 is required for steps in the normal medial and trans Golgi-associated processing of glycoconjugates. It plays a role in the organization of the Golgi-localized complex. Mutations in COG1 in humans cause novel types of congenital disorders of glycosylation (CDG). COG complex regulates membrane trafficking and maintenance of Golgi glycosylation machinery in eukaryotic cells.
Conserved oligomeric Golgi (COG) complex plays a vital role in retrograde vesicular trafficking and glycosylation.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cerebrocostomandibular-like syndrome and a mutation in the conserved oligomeric Golgi complex, subunit 1.
Zeevaert R
Human Molecular Genetics, 18, 517-524 (2009)
Genome-wide Examination of Chromosomal Aberrations in Neuroblastoma SH-SY5Y Cells by Array-based Comparative Genomic Hybridization
Do JH
Molecules and Cells, 24, 105-112 (2007)
Role of the conserved oligomeric Golgi (COG) complex in protein glycosylation
Smith RD and Lupashin VV
Carbohydrate Research, 343(12), 2024-2031 (2008)
Conserved oligomeric Golgi complex specifically regulates the maintenance of Golgi glycosylation machinery
Pokrovskaya ID, et al.
Glycobiology, 21(12), 1554-1569 (2011)
Comparative analyses of the Conserved Oligomeric Golgi (COG) complex in vertebrates
Quental R, et al.
BMC Evolutionary Biology, 10(1), 212-212 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service