Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB4200340

Sigma-Aldrich

Anti-Golph3 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-GOPP1, Anti-GPP34, Anti-Golgi peripheral membrane protein 1, 34 kDa, Anti-Golgi phosphoprotein 3 (coat-protein), Anti-Golgi-associated protein, Anti-MIDAS, Anti-Mitochondrial DNA absence factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human HeLa cells.
western blot: 0.1-0.2 μg/mL using whole extracts of HEK-293 cells over-expressing human Golph3.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GOLPH3(64083)
mouse ... Golph3(66629)

General description

Golgi phosphoprotein 3 (Golph3) is a peripheral membrane protein of the Golgi stack that localizes to the trans-Golgi network. It is conserved from yeast to human. The GOLPH3 gene is mapped to the human chromosome 5p13.3 location.

Specificity

Anti-Golph3 (C-terminal) recognizes human Golph3.

Immunogen

peptide corresponding to the C-terminal region of human Golph3, conjugated to KLH. The corresponding sequence is identical in mouse, rat, monkey and canine.

Application

Anti-Golph3 (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Golgi phosphoprotein 3 (Golph3) is a phosphatidylinositol-4-phosphate (PtdIns(4)P) binding protein required for Golgi function. It plays a vital role in Golgi trafficking and morphology by interacting with the unconventional myosin 18A (MYO18A) linking Golgi membranes to the F-actin cytoskeleton. GOLPH3 is an oncogene that is commonly targeted for amplification in human cancer and cancer cell lines. Furthermore, Golph3 overexpression is correlated with the hyperactivation of the mammalian target of rapamycin (mTOR) signaling in human cells.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The multiple cellular functions of the oncoprotein Golgi phosphoprotein 3
Sechi S, et al.
Oncotarget, 6(6), 3493-3493 (2015)
De novo 15.5-Mb Interstitial Deletion in 5p in a Male Ascertained by Oligospermia
Papoulidis I, et al.
Molecular Syndromology, 4(5), 250-254 (2013)
Study of GOLPH3: a potential stress-inducible protein from Golgi apparatus
Li T, et al.
Molecular Neurobiology, 49(3), 1449-1459 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service