Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200276

Sigma-Aldrich

Anti-RCC1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CHC-1, Anti-CHC1, Anti-Chromosome Condensation 1, Anti-Regulator of Chromatin Condensation 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using lysates of HeLa or HEK-293T cells.
indirect immunofluorescence: 1-2 μg/mL using paraformaldehyde fixed HeLa cells.
western blot: 2-4 μg/mL using lysates of HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RCC1(1104)
mouse ... Rcc1(100088)
rat ... Rcc1(682908)

Immunogen

synthetic peptide corresponding to a sequence at the C- terminal region of human RCC1, conjugated to KLH. The corresponding sequence is identical in rat and mouse

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service