Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200242

Sigma-Aldrich

Anti-MAN1C1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-1,2-alpha-mannosidase IC, mannosyl-oligosaccharide 1,2-alpha-mannosidase IC, Anti-HMIC, Anti-MAN1A3, Anti-MAN1C, Anti-mannosidase, alpha, class 1C, member 1, Anti-pp6318, Anti-processing alpha-1,2-mannosidase IC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~71 kDa

species reactivity

rat, human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-0.2 μg/mL using whole extracts of HEK-293 cells overexpressing human MAN1C1
western blot: 5-10 μg/mL using whole extracts of rat liver

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Mannosidase alpha class 1C member 1 (MAN1C1) is a member of the glycosyl hydrolase 47 family. MAN1C1 is a type II membrane protein with sequence similarity to class Iα,2-mannosidases, which localizes to the Golgi apparatus membrane.

Application

Anti-MAN1C1 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Mannosidase alpha class 1C member 1 (MAN1C1) is involved in the maturation of Asn-linked oligosaccharides and quality control. It trims α-1,2-linked mannose residues from Glc3Man9GlcNAc2 to produce the Man5GlcNAc2 processing intermediate. MAN1C1 overexpression accelerates endoplasmic reticulum-associated protein degradation (ERAD) of terminally misfolded human null Hong Kongα1-antitrypsin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a cDNA encoding a novel human Golgi alpha1, 2-mannosidase (IC) involved in N-glycan biosynthesis
Tremblay LO and Herscovics A
The Journal of Biological Chemistry, 275(41), 31655-31660 (2000)
Family 47 alpha-mannosidases in N-glycan processing
Mast SW and Moremen KW
Test, 415, 31-46 (2006)
Nobuko Hosokawa et al.
Biochemical and biophysical research communications, 362(3), 626-632 (2007-08-31)
Terminally misfolded or unassembled proteins are degraded by the cytoplasmic ubiquitin-proteasome pathway in a process known as ERAD (endoplasmic reticulum-associated protein degradation). Overexpression of ER alpha1,2-mannosidase I and EDEMs target misfolded glycoproteins for ERAD, most likely due to trimming of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service