Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200218

Sigma-Aldrich

Anti-Glycogen Synthase 1 (N-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-GYS1, Anti-Glycogen synthase 1 (muscle)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~84 kDa

species reactivity

mouse, rat

concentration

~1.5 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using rat brain extracts (S1 fraction)
indirect immunofluorescence: 5-10 μg/mL using NIH3T3 cells
western blot: 1.5-3 μg/mL using mouse brain extracts (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GYS1(2997)

General description

The glycogen synthase 1 (GYS1) gene is mapped to human chromosome 19q13.3. In mammals, two glycogen synthase (GS) isoforms have been identified, the muscle GS form, glycogen synthase 1 (GYS1) and the liver specific form, glycogen synthase 2 (GYS2, LGS). GYS1 is predominantly expressed in the skeletal and cardiac muscles.

Specificity

Anti-Glycogen Synthase 1 (N-terminal), specifically recognizes mouse and rat glycogen synthase 1.

Application

Anti-Glycogen Synthase 1 (N-terminal) antibody produced in rabbit is suitable for:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Glycogen synthase (GS or GYS), the rate-limiting enzyme for glycogen biosynthesis, catalyzes the incorporation of α-1,4-linked glucose units into glycogen chains. Both, the muscle (GYS1) and the liver GS (GYS2) forms are highly regulated by phosphorylation, glucose availability, glycogen levels and allosteric effectors. In response to hormonal stimuli, GS is phosphorylated on up to nine serine residues resulting in progressive inactivation of the two isoforms. GS activity is stimulated by insulin in liver, muscle and adipose tissues. The enzyme undergoes dephosphorylation allosteric activation and thus translocates between various subcellular compartments. Insulin increases GS activity primarily by dephosphorylation of four key residues, a process thought to be mediated by the activation of protein phosphatase-1 (PP1) and the inactivation of glycogen synthase kinase-3 (GSK3). GYS1 deficiency might lead to congenital disorders associated with glycogen metabolism.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T C Jensen et al.
The Journal of biological chemistry, 275(51), 40148-40154 (2000-10-03)
A protocol was developed in 3T3-L1 adipocytes that resulted in the specific desensitization of glycogen synthase activation by insulin. Cells were pretreated for 15 min with 100 nm insulin, and then recovered for 1.5 h in the absence of hormone.
Juan C Ferrer et al.
FEBS letters, 546(1), 127-132 (2003-06-28)
Traditionally, glycogen synthase (GS) has been considered to catalyze the key step of glycogen synthesis and to exercise most of the control over this metabolic pathway. However, recent advances have shown that other factors must be considered. Moreover, the control
María M Adeva-Andany et al.
BBA clinical, 5, 85-100 (2016-04-07)
In the human body, glycogen is a branched polymer of glucose stored mainly in the liver and the skeletal muscle that supplies glucose to the blood stream during fasting periods and to the muscle cells during muscle contraction. Glycogen has
Alexander von Wilamowitz-Moellendorff et al.
Diabetes, 62(12), 4070-4082 (2013-08-31)
The liver responds to an increase in blood glucose levels in the postprandial state by uptake of glucose and conversion to glycogen. Liver glycogen synthase (GYS2), a key enzyme in glycogen synthesis, is controlled by a complex interplay between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service