Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200212

Sigma-Aldrich

Anti-Nedd9 antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CAS-L, Anti-CAS2, Anti-CASL (CRK-Associated substrate-related protein), Anti-CASS2, Anti-HEF1 (Human Enhancer of Filamentation 1), Anti-Neural precursor cell expressed, developmentally downregulated 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~115 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 2-4 μg/mL using HS68 cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEDD9(4739)

General description

Neural precursor cell expressed developmentally down-regulated protein 9 (Nedd9), also known as CRK-associated substrate-related protein (Cas-L) and human enhancer of filamentation 1 (HEF1), is an adhesion adapter protein. Nedd9 gene is mapped to human chromosome 6p24.2. It belongs to a larger family of Cas docking adapter proteins and is expressed preferentially in lymphocytes and epithelial cells.

Specificity

Anti-Nedd9 specifically recognizes human Nedd9.

Application

Anti-Nedd9 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Neural precursor cell expressed developmentally down-regulated protein 9 (Nedd9) participates in integrin and growth factor signaling pathways that regulate growth, motility and apoptosis. It is thought to play an important role in early neuronal differentiation and brain development. Nedd9 undergoes extensive phosphorylation, a process dependent both on cell adhesion and on the integrity of the actin cytoskeleton. Focal adhesion kinase (FAK) and sarcoma (Src) regulate Nedd9. Nedd9 is implicated in metastatic cancer and is upregulated in solid tumors and ovarian cancer. Overexpression of Nedd9 is implicated in the invasion of glioblastoma and in the pathogenesis of breakpoint cluster region-Abelson (Bcr-Abl)-dependent tumor .

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua-Xin Duan et al.
American journal of physiology. Gastrointestinal and liver physiology, 316(6), G763-G773 (2019-03-29)
Tumor-associated angiogenesis plays a critical role in the pathogenesis of cholangiocarcinoma (CCA). In this study, we examined the biological effects and molecular mechanisms of transcription factor 21 (TCF21) on CCA-associated angiogenesis. TCF21 expression was compared between 15 pairs of peritumor
Roles of neural precursor cell expressed, developmentally downregulated 9 in tumor-associated cellular processes
Zhang S and Wu L
Molecular Medicine Reports, 12(5), 6415-6421 (2015)
Christian Dölle et al.
Scientific reports, 8(1), 1272-1272 (2018-01-21)
3,3'-Diaminobenzidine (DAB) is a widely used chromogen in histological staining methods and stained tissue is often used in downstream molecular analyses such as quantitative PCR (qPCR). Using microdissected muscle fibers from sequential muscle sections stained by DAB-dependent and -independent methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service