Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200059

Sigma-Aldrich

Anti-Chimerin β2 antibody, Rat monoclonal

clone HAR 2E3, purified from hybridoma cell culture

Synonym(s):

Anti-ARHGAP3, Anti-BCH, Anti-CHN2, Anti-Chimaerin β2, Anti-GTPase-activating protein, RHO, 3, Anti-RHOGAP3, Anti-Rho-GTPase-activating protein 3, Monoclonal Anti-Chimerin β2 antibody produced in rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HAR 2E3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human, monkey, rat, mouse, bovine

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: 1.0-2.0 μg/mL using Neuro-2a cell extracts

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHN2(1124)

General description

Monoclonal Anti-Chimerin β2 (rat IgG2a isotype) is derived from the hybridoma HAR 2E3 produced by the fusion of mouse myeloma cells and splenocytes from rat immunized with a fusion protein of full length human Chimerin β2. Chimerin β2 (CHN2) is a member of the chimerin family and encodes a protein with a phorbol-ester/ diacylglycerol (DAG)-type zinc finger containing C1 domain, a Rho-GTPase activating proteins (GAP) domain and an Src homology 2 (SH2) domain. Chimerin β2 gene is mapped to human chromosome 7p14.3.

Specificity

Monoclonal Anti-Chimerin b2 recognizes human, monkey, bovine, mouse and rat Chimerin b2.

Immunogen

fusion protein of full length human Chimerin β2

Application

Monoclonal Anti-Chimerin β2 antibody produced in rat has been used in immunoblotting at a dilution of 1:1000 and immunocytochemistry.

Biochem/physiol Actions

Chimerin β2 protein (CHN2) is a high affinity receptor for the phorbol esters and diacylglycerol (DAG). Binding of DAG induces translocation of the protein from the cytosol to the Golgi apparatus membrane. It has guanosine triphosphatase GTPase-activating protein activity regulated by phospholipid binding. Decreased expression of CHN2 gene is associated with high-grade gliomas and breast. Deregulation in expression of CHN2 gene is associated with lymphomas, glioblastoma and insulin resistance. Mutations in the CHN2 gene have been associated with schizophrenia in men.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at -20 °C. For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing,, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcus G Pezzolesi et al.
Kidney international, 80(1), 105-111 (2011-03-18)
A genome-wide association scan of the Genetics of Kidneys in Diabetes (GoKinD) collections identified four novel susceptibility loci, located on chromosomes 7p14.3, 9q21.32, 11p15.4, and 13q33.3 associated with type 1 diabetic nephropathy. A recent evaluation of these loci in Japanese
A missense polymorphism (H204R) of a Rho GTPase-activating protein, the chimerin 2 gene, is associated with schizophrenia in men.
Ryota Hashimoto et al.
Schizophrenia research, 73(2-3), 383-385 (2005-01-18)
M J Caloca et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(21), 11854-11859 (1999-10-16)
The members of the chimaerin family of Rac-GTPase-activating proteins possess a single C1 domain with high homology to those present in protein kinase C (PKC) isozymes. This domain in PKCs is involved in phorbol ester and diacylglycerol (DAG) binding. We
Victoria Casado-Medrano et al.
Oncotarget, 7(19), 28301-28319 (2016-04-09)
β2-chimaerin is a Rac1-specific negative regulator and a candidate tumor suppressor in breast cancer but its precise function in mammary tumorigenesis in vivo is unknown. Here, we study for the first time the role of β2-chimaerin in breast cancer using
Martin J Baker et al.
Cancers, 12(2) (2020-02-26)
The GTPase Rac1 is a well-established master regulator of cell motility and invasiveness contributing to cancer metastasis. Dysregulation of the Rac1 signaling pathway, resulting in elevated motile and invasive potential, has been reported in multiple cancers. However, there are limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service