Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4100327

Sigma-Aldrich

Monoclonal Anti-ADIPOQ antibody produced in mouse

clone ADIPOQ-A277, culture supernatant

Synonym(s):

Anti-ACDC, Anti-ACRP30, Anti-ADIPQTL1, Anti-ADPN, Anti-APM-1, Anti-APM1, Anti-Adiponectin, C1Q and collagen domain containing, Anti-GBP28

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

ADIPOQ-A277, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 24.9 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ADIPOQ(9370)

General description

Adiponectin (ADIPOQ) gene is mapped to human chromosome 3q27.3. The gene codes for a 244 amino acid protein with four distinct regions. ADIPOQ is secreted by the adipocytes and is a most abundant circulating hormone.[1]

Immunogen

ADIPOQ purified recombinant protein

Biochem/physiol Actions

Adiponectin (ADIPOQ) is an adipocyte specific secreted protein that circulates in the plasma.[2] The protein has a C-terminal globular domain and an NH2-terminal collagen domain.[3] It has anti-atherosclerotic and anti-inflammatory properties.[4] ADIPOQ has a role in physiological processes such as energy homeostasis and obesity. [5] It exhibits anti-diabetic action by interacting with adiponectin receptors, AdipoR1 and AdipoR2.[3] Low levels of adiponectin are also linked with hypertension and dyslipidemia.[5] Mutation in the gene encoding it is linked with risk for endometrial cancer.[6]

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic architecture of the APM1 gene and its influence on adiponectin plasma levels and parameters of the metabolic syndrome in 1,727 healthy Caucasians
Heid IM, et al.
Diabetes, 55, 375-384 (2019)
Association Between Single Nucleotide Polymorphism +276G T (rs1501299) in ADIPOQ and Endometrial Cancer
Bienkiewicz J, et al.
Pathology Oncology Research, 22, 135-138 (2019)
Adiponectin and adiponectin receptors in insulin resistance, diabetes, and the metabolic syndrome
Kadowaki T, et al.
The Journal of Clinical Investigation, 116, 1784-1792 (2019)
In silico Evaluation of Nonsynonymous Single Nucleotide Polymorphisms in the ADIPOQ Gene Associated with Diabetes, Obesity, and Inflammation
Swamy AN, et al.
Avicenna Journal of Medical Biotechnology, 7(6), 121-121 (2015)
Plasma Concentrations of a Novel, Adipose-Specific Protein, Adiponectin, in Type 2 Diabetic Patients
Hotta K, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service