Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3700518

Sigma-Aldrich

Anti-Rat IgA (α-chain specific) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Rat IgA coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Goat Serum, Rat IgA and Rat Serum. No reaction was observed against Rat IgM or Rat IgG. Specificity was confirmed by ELISA at less than 1% cross reactivity against other Rat heavy or light chain isotypes.

Immunogen

Rat IgA alpha heavy chain

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.1 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah L Heyl et al.
FEBS letters, 593(7), 732-742 (2019-03-02)
Dysfunction of the dopaminergic pathway is linked to numerous diseases of the nervous system. The D1-D2 receptor heteromer is known to play a role in certain neuropsychiatric disorders, such as depression. Here, we synthesized an eight amino acid residue peptide
Deborah L Heyl et al.
FEBS letters, 593(7), 732-742 (2019-03-02)
Dysfunction of the dopaminergic pathway is linked to numerous diseases of the nervous system. The D1-D2 receptor heteromer is known to play a role in certain neuropsychiatric disorders, such as depression. Here, we synthesized an eight amino acid residue peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service