Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB3501258

Sigma-Aldrich

Anti-SFTS Virus HB29 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (SFTS virus HB29)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

SFTS Virus HB29 Antibody: Severe fever with thrombocytopenia syndrome (SFTS) is an emerging infectious disease caused by SFTS virus, a newly discovered bunyavirus that can cause high rates of fatalities. This disease is thought to be transmitted through blood contact. The SFTS virus HB29 membrane glycoprotein polyprotein mRNA encodes two glycoproteins termed Phlebovirus glycoprotein G1 and G2 respectively. This antibody will detect the G1 glycoprotein.

Immunogen

SFTS Virus HB29 Membrane Glycoprotein antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SFTS Virus HB29 Membrane Glycoprotein.The immunogen is located within amino acids 60 - 110 of SFTS Virus HB29.

Application

SFTS Virus HB29 Membrane Glycoprotein antibody can be used for detection of SFTS Virus HB29 Membrane Glycoprotein by ELISA. It will detect 10 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service