Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3501201

Sigma-Aldrich

Anti-Anthrax Edema Factor antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$603.00

$603.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$603.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

Bacillus anthracis

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anthrax Edema Factor Antibody: Anthrax infection is initiated by the inhalation, ingestion, or cutaneous contact with Bacillus anthracis endospores. B. anthracis produces three polypeptides that comprise the anthrax toxin: protective antigen (PA), lethal factor (LF), and edema factor (EF). PA binds to two related proteins on the cell surface; these are termed tumor epithelial marker 8 (TEM8)/anthrax toxin receptor (ATR) and capillary morphogenesis protein 2 (CMG2), although it is still unclear which is physiologically relevant. Following PA binding to its receptor, PA is cleaved into two fragments by a furin-like protease. The bound fragment binds both LF and EF; the resulting complex is then endocytosed which allows the translocation of LF and EF into the cytoplasm. EF is a calmodulin and Ca++-dependent adenylate cyclase responsible for the edema seen in the disease. It is thought to benefit the B. anthracis bacteria by inhibiting cells of the host immune system.

Immunogen

Anthrax Edema Factor antibody was raised against a synthetic peptide corresponding to 16 amino acids near the carboxy terminus of the Anthrax edema factor protein.The immunogen is located within the last 50 amino acids of Anthrax Edema Factor.

Application

Anthrax Edema Factor antibody can be used for the detection of Anthrax PA protein in ELISA. It will detect 10 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service