Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB3500414

Sigma-Aldrich

Anti-IKKγ antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FIP3, Anti-IKKg / NEMO, Anti-NEMO

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$540.00

$540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$540.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 52 kDa

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IKBKG(8517)

Immunogen

IKK gamma antibody was raised against a peptide corresponding to amino acids 400 to 416 of human IKK gamma, which are identical to those of mouse homologue.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500414.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beibei Fu et al.
Cell death & disease, 11(9), 803-803 (2020-09-27)
Acute liver failure (ALF) is a rare but life-threatening systemic disorder. The innate immune regulation has an important role in this process; however, the specific mechanisms are not completely clear. Using the LPS + D-GalN-induced ALF mouse model, we found that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service