Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB3500026

Sigma-Aldrich

Anti-ATOH8 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HATH6, Anti-Helix-loop-helix protein hATH-6, Anti-MATH6, Anti-Protein atonal homolog 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$506.00

$506.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$506.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$506.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 35 kDa

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATOH8(84913)

Immunogen

ATOH8 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human ATOH8.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Basic helix-loop-helix (bHLH) transcription factors play important roles in differentiation processes during embryonic development of vertebrates. ATOH8 (MATH6) is a tissue-restricted member of the atonal superfamily of bHLH transcription factors that exhibits 43–57% identity in the bHLH domain with other mammalian atonal paralogs including the NeuroD and Neurogenin factors. In the mouse, ATOH8 has been implicated in the specification and differentiation of neuronal cell lineages in the brain and may also participate in kidney development. Recent studies show that ATOH8 is a novel component of the pancreatic transcriptional network during embryonic development and suggest a potential role as a modulator of the differentiation program initiated by the pro-endocrine factor Neurog3. It is indispensable for early embryonic development, suggesting a more widespread function for this factor in tissue-specific differentiation processes that are dependent on class II bHLH genes.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500026.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanxiu Li et al.
Journal of cellular physiology, 234(11), 21307-21315 (2019-05-09)
Retinal diseases are characterized by the degeneration of retinal neural cells, and are the main cause of blindness. Although the development of stem cell including retinal stem cell therapies raises hope for retinal neuron replacement, currently, there is still no
Yanxiu Li et al.
Journal of cellular physiology, 234(11), 21307-21315 (2019-05-09)
Retinal diseases are characterized by the degeneration of retinal neural cells, and are the main cause of blindness. Although the development of stem cell including retinal stem cell therapies raises hope for retinal neuron replacement, currently, there is still no

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service