Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB3300047

Sigma-Aldrich

Monoclonal Anti-TUBB3 antibody produced in mouse

clone 5H2, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Anti-β-4, Anti-MC1R, Anti-TUBB4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H2, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen 50.3 kDa

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TUBB3(10381)

Immunogen

Recombinant protein expressed in E. coli corresponding to amino acids 1-150 of human TUBB3

Target description

Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and the cytoskeleton. Except in the simplest eukaryotes, tubulin exists in all cells as a mixture of similar, but not identical, sets of alpha and beta tubulin polypeptides. Within either set of polypeptides, individual subunits diverge from each other (both within and across species) at less than 10% of the amino acid positions. The most extreme diversity is localized to the 15 residues of the carboxy terminal. For β-tubulin five evolutionarily conserved isotype clones have been identified. These are almost totally conserved in the subunits utilized in the same cell types of different species, with the exception of the hematopoietic β-tubulin which is the most highly divergent in sequence and is not conserved between species. Research has been centered around the hypothesis that these beta tubulin isotypes contribute to unique functional properties. It has been reported that the different isotypes of tubulin differ from each other in their ability to polymerize into microtubules.

Physical form

Solution in phosphate buffered saline containing 1% bovine serum albumin, 50% glycerol, and 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Palmero et al.
Scientific reports, 6, 39394-39394 (2016-12-20)
In our lab we study neurogenesis and the development of brain tumors. We work towards treatment strategies for glioblastoma and towards using autologous neural stem cells for tissue regeneration strategies for brain damage and neurodegenerative disorders. It has been our
Jessica E M Dunleavy et al.
Development (Cambridge, England), 148(24) (2021-11-26)
Katanin microtubule-severing enzymes are crucial executers of microtubule regulation. Here, we have created an allelic loss-of-function series of the katanin regulatory B-subunit KATNB1 in mice. We reveal that KATNB1 is the master regulator of all katanin enzymatic A-subunits during mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service