Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2702279

Sigma-Aldrich

Monoclonal Anti-Influenza B Virus NP antibody produced in mouse

clone GT371, affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$666.00

$666.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$666.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$666.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT371, monoclonal

form

buffered aqueous solution

species reactivity

Influenza B virus

concentration

1mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 1000-10000

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Recombinant fragment corresponding to N-terminal of Influenza B virus NP (B/Taiwan/753/2005)

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:2000, WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service