Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB2702254

Sigma-Aldrich

Monoclonal Anti-Influenza A virus NP (nucleoprotein) antibody produced in mouse

clone GT1236, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT1236, monoclonal

form

buffered aqueous solution

mol wt

56kDa

species reactivity

influenza A virus

technique(s)

immunofluorescence: 1:100-1:1,000
immunohistochemistry: suitable
western blot: 1:1,000-1:10,000

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Recombinant fragment contain a sequence corresponding to a region within amino acids 1 and 498 of Influenza A Virus H1N1 NP

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service