Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

SAB2701964

Sigma-Aldrich

Anti-HSPA8 antibody produced in rabbit

Synonym(s):

HSP73, HSPA10, LAP1, NIP71

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

71 kDa

species reactivity

rat, human, mouse

concentration

10 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1000-20000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HSPA8(3312)

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 470 and 635 (P11142) of Hsc70

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:1000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

The product encoded by this gene belongs to the heat shock protein 70 family which contains both heat-inducible and constitutively expressed members. The latter are called heat-shock cognate proteins. This gene encodes a heat-shock cognate protein. This protein binds to nascent polypeptides to facilitate correct folding. It also functions as an ATPase in the disassembly of clathrin-coated vesicles during transport of membrane components through the cell. Two alternatively spliced variants have been characterized to date. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed I Abdelwahab Hassan et al.
Environmental microbiology, 22(9), 3722-3740 (2020-06-26)
Mucormycosis is an emergent, fatal fungal infection of humans and warm-blooded animals caused by species of the order Mucorales. Immune cells of the innate immune system serve as the first line of defence against inhaled spores. Alveolar macrophages were challenged
Hanghui Wang et al.
International journal of oncology, 45(1), 157-164 (2014-04-17)
The specific and efficient delivery of small interfering RNA (siRNA) into cancer cells in vivo remains a major obstacle. In this study, we investigated whether ultrasound-targeted microbubble destruction (UTMD) combined with dual targeting of HSP72 and HSC70 in prostate cancer cell
Nida S Iqbal et al.
BioTechniques, 56(5), 239-249 (2014-05-09)
Although many researchers have successfully uncovered novel functions of the tumor suppressor p19(Arf) utilizing various types of cultured cancer cells and immortalized fibroblasts, these systems do not accurately reflect the endogenous environment in which Arf is developmentally expressed. We addressed
Thierry Langlois et al.
Stem cells (Dayton, Ohio), 32(8), 2084-2097 (2014-04-12)
Ten-eleven-translocation 2 (TET2) belongs to the TET protein family that catalyzes the conversion of 5-methylcytosine into 5-hydroxymethylcytosine and plays a central role in normal and malignant adult hematopoiesis. Yet the role of TET2 in human hematopoietic development remains largely unknown.
Najmeh Moein-Vaziri et al.
Reproduction (Cambridge, England), 147(5), 719-732 (2014-02-07)
The constitutive 70  kDa heat-shock protein, HSPA8, has previously been shown to contribute to the long-term survival of spermatozoa inside the mammalian female reproductive tract. Here, we show that a recombinant form of HSPA8 rapidly promotes the viability of uncapacitated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service