Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501926

Sigma-Aldrich

Anti-IDO1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CD107B, HGNC:6059, IDO, INDO, indoleamine-pyrrole 2,3 dioxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IDO1(3620)

General description

Indoleamine 2,3 dioxygenase (IDO) 1 is one of the four tryptophan catabolic enzymes in mammals. It codes for a full-length protein, made up of 403 amino acids and has a molecular weight of 45,332 Daltons. It is present in the cytosol. IDO1 is located on human chromosome 8p11.

Immunogen

Peptide with sequence CVLANWKKKDPNK, from the internal region of the protein sequence according to NP_002155.1

Biochem/physiol Actions

Indoleamine 2,3 dioxygenase (IDO) 1 plays an important role in the metabolism of tryptophan and in immune regulation. Removal of IDO1 results in attenuated contact hypersensitivity (CHS) responses.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IDO1 (indoleamine 2, 3-dioxygenase 1)
Chang Mee Young, et al.
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2010)
Indoleamine-2, 3-dioxygenase (IDO) 2 polymorphisms are not associated with multiple sclerosis in Italians
C Agliardi, et al.
Journal of the Neurological Sciences, 377, 31-34 (2017)
Richard Metz et al.
International immunology, 26(7), 357-367 (2014-01-10)
IDO2 is implicated in tryptophan catabolism and immunity but its physiological functions are not well established. Here we report the characterization of mice genetically deficient in IDO2, which develop normally but exhibit defects in IDO-mediated T-cell regulation and inflammatory responses.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service