Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501804

Sigma-Aldrich

Anti-CCL20 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CKb4, LARC, MIP-3a, chemokine (C-C motif) ligand 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, bovine, pig

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL20(6364)

General description

Chemokine ligand 20 (CCL20) is also called as liver and activation-regulated chemokine, macrophage inflammatory protein-3α (MIP-3α) or exodus-1. It is a cysteine−cysteine chemokine, that is located on human chromosome 2q36. It is expressed at high levels in serum or IgA1 and HMC incubation system. The CCL20 promoter region consists of a transcription start site, a nuclear factor (NF)-κB binding site, a CCAAT/enhancer-binding proteins binding site, an activator protein-1 binding sites and a specificity protein 1 (Sp1)-binding site.

Immunogen

Peptide with sequence CLGYTDRILHPK, from the internal region of the protein sequence according toNP_004582.1; NP_001123518.1

Biochem/physiol Actions

Chemokine ligand 20 (CCL20) is chemotactic for immature dendritic cells, effector or memory CD4+T lymphocytes and B lymphocytes. CCR6 (C-C chemokine receptor type 6), the receptor of CCL20, plays a vital role in the progression of several diseases, such as tumors, rheumatoid arthritis, pneumococcal meningitis and human renal inflammation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide linkage scan of a large family with IgA nephropathy localizes a novel susceptibility locus to chromosome 2q36
A D Paterson, et al.
Journal of the American Society of Nephrology, 18(8), 2408-2415 (2007)
Transcriptional regulation of CCL20 expression
Zhao Lifang, et al.
Microbes and Infection, 16(10), 864-870 (2014)
Richard Metz et al.
International immunology, 26(7), 357-367 (2014-01-10)
IDO2 is implicated in tryptophan catabolism and immunity but its physiological functions are not well established. Here we report the characterization of mice genetically deficient in IDO2, which develop normally but exhibit defects in IDO-mediated T-cell regulation and inflammatory responses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service