Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2500606

Sigma-Aldrich

Anti-LXR α/LXR β antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NR1H3, Anti-RLD-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$637.00

$637.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$637.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$637.00


Please contact Customer Service for Availability

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Liver X receptor α and β (LXR-α/LXR-β) are members of LXRs that belong to the nuclear receptor superfamily. LXR-α is expressed predominantly in macrophages, liver, intestine, kidney, lung and adipose tissue. The protein contains a DNA-binding domain that requires the ligand-binding domain to heterodimerize with a retinoid X receptor partner for its DNA-binding activity. The gene encoding this protein is localized on to human chromosome 11p11.2.

Immunogen

Peptide with sequence CRLQDKKLPPLLSEI from the internal region of the protein sequence according to NP_005684; NP_009052.

Biochem/physiol Actions

Liver X receptor α (LXR-α) has an important role in the control of cholesterol homeostasis. LXRs have been found to regulate inflammatory response in macrophages. They are also involved in the regulation of cell proliferation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CCAR2 negatively regulates nuclear receptor LXRa by competing with SIRT1 deacetylase.
Sakurabashi A
The Journal of Steroid Biochemistry and Molecular Biology (2015)
Liver x receptors in atherosclerosis and inflammation.
Im SS and Osborne TF
Circulation Research (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service