Skip to Content
MilliporeSigma
All Photos(3)

Documents

SAB2108245

Sigma-Aldrich

Anti-TRPC4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HTRP-4, Anti-HTRP4, Anti-TRP4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

112kDa

species reactivity

mouse, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPC4(7223)

Immunogen

Synthetic peptide directed towards the middle region of human TRPC4

Biochem/physiol Actions

TRPC4 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC4 probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. It has also been shown to be calcium-selective. TRPC4 may also be activated by intracellular calcium store depletion.

Sequence

Synthetic peptide located within the following region: CPFKSEKVVVEDTVPIIPKEKHAKEEDSSIDYDLNLPDTVTHEDYVTTRL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Conte et al.
Frontiers in pharmacology, 15, 1393746-1393746 (2024-07-04)
Introduction: During aging, sarcopenia and decline in physiological processes lead to partial loss of muscle strength, atrophy, and increased fatigability. Muscle changes may be related to a reduced intake of essential amino acids playing a role in proteostasis. We have
Tamas Papp et al.
Frontiers in neuroscience, 16, 803356-803356 (2022-04-05)
Neuronal differentiation and synaptogenesis are regulated by precise orchestration of intrinsic and extrinsic chemical and mechanical factors throughout all developmental steps critical for the assembly of neurons into functional circuits. While ultrasound is known to alter neuronal migration and activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service